Assay Details
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174848.3 | NP_777273.2 | 15 - 16 | 3193 | 116 | |
GenBank mRNA | AK030282.1 | - | 6 - 7 | 637 | 116 | |
AK162506.1 | - | 3 - 4 | 1028 | 116 | ||
BC043118.1 | - | 15 - 16 | 3193 | 6408859 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
