Assay Details
| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081190.1 | NP_001074659.1 | 1 - 2 | 117 | 69 | |
| GenBank mRNA | BC151105.1 | - | 1 - 2 | 167 | 9088018 | 69 |
More Information
Updated Mapping Information
Set Membership: |
|

Gabrr3
Genomic Map