Assay Details
| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081190.1 | NP_001074659.1 | 2 - 3 | 231 | 89 | |
| GenBank mRNA | BC151105.1 | - | 2 - 3 | 281 | 9088018 | 89 |
More Information
Set Membership: |
|

Gabrr3
Genomic Map