Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019827.6 | NP_062801.1 | 1 - 2 | 1609 | 83 | |
| GenBank mRNA | AB066114.1 | - | 1 - 2 | 1608 | 83 | |
| AF156099.2 | - | 1 - 2 | 250 | 83 | ||
| AK154293.1 | - | 1 - 2 | 1087 | 83 | ||
| AK170965.1 | - | 1 - 2 | 1056 | 83 | ||
| AK171482.1 | - | 1 - 2 | 1126 | 83 | ||
| AK190946.1 | - | 1 - 2 | 53 | 83 | ||
| BC006936.1 | - | 1 - 2 | 116 | 2648507 | 83 | |
| BC060743.1 | - | 1 - 2 | 1536 | 4022374 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsk3b
Genomic Map