Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019827.6 | NP_062801.1 | 2 - 3 | 1807 | 90 | |
| GenBank mRNA | AB066114.1 | - | 2 - 3 | 1806 | 90 | |
| AF156099.2 | - | 2 - 3 | 448 | 90 | ||
| AK154293.1 | - | 2 - 3 | 1285 | 90 | ||
| AK164048.1 | - | 1 - 2 | 159 | 90 | ||
| AK170965.1 | - | 2 - 3 | 1254 | 90 | ||
| AK190946.1 | - | 2 - 3 | 251 | 90 | ||
| BC006936.1 | - | 2 - 3 | 314 | 2648507 | 90 | |
| BC060743.1 | - | 2 - 3 | 1734 | 4022374 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsk3b
Genomic Map