Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019827.6 | NP_062801.1 | 3 - 4 | 1898 | 91 | |
| GenBank mRNA | AB066114.1 | - | 3 - 5 | 1897 | 91 | |
| AF156099.2 | - | 3 - 4 | 539 | 91 | ||
| AK154293.1 | - | 3 - 5 | 1376 | 91 | ||
| AK164048.1 | - | 2 - 3 | 250 | 91 | ||
| AK170965.1 | - | 3 - 5 | 1345 | 91 | ||
| BC006936.1 | - | 3 - 4 | 405 | 2648507 | 91 | |
| BC060743.1 | - | 3 - 5 | 1825 | 4022374 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsk3b
Genomic Map