Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019827.6 | NP_062801.1 | 5 - 6 | 2135 | 120 | |
| GenBank mRNA | AB066114.1 | - | 6 - 7 | 2134 | 120 | |
| AF156099.2 | - | 5 - 6 | 776 | 120 | ||
| AK154293.1 | - | 6 - 7 | 1613 | 120 | ||
| AK164048.1 | - | 4 - 5 | 487 | 120 | ||
| AK170965.1 | - | 6 - 7 | 1582 | 120 | ||
| BC006936.1 | - | 5 - 6 | 642 | 2648507 | 120 | |
| BC060743.1 | - | 6 - 7 | 2062 | 4022374 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsk3b
Genomic Map