Assay Details
| Gene Symbol: Gsk3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019827.6 | NP_062801.1 | 9 - 10 | 2624 | 81 | |
| GenBank mRNA | AB066114.1 | - | 10 - 11 | 2623 | 81 | |
| AF156099.2 | - | 9 - 10 | 1265 | 81 | ||
| AK154293.1 | - | 10 - 11 | 2102 | 81 | ||
| AK170965.1 | - | 10 - 11 | 2071 | 81 | ||
| BC006936.1 | - | 9 - 10 | 1131 | 2648507 | 81 | |
| BC060743.1 | - | 10 - 11 | 2551 | 4022374 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gsk3b
Genomic Map