Assay Details
| Gene Symbol: Nr1i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098404.1 | NP_001091874.1 | 7 - 8 | 1257 | 92 | |
| NM_010936.3 | NP_035066.1 | 7 - 8 | 1380 | 92 | ||
| GenBank mRNA | AF031814.1 | - | 7 - 8 | 1195 | 92 | |
| AK018630.1 | - | 7 - 8 | 1380 | 92 | ||
| BC120796.1 | - | 7 - 8 | 1127 | 40129719 | 92 | |
| BC137780.1 | - | 7 - 8 | 1127 | 8860799 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr1i2
Genomic Map