Assay Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300827.1 | NP_001287756.1 | 2 - 3 | 470 | 69 | |
| NM_172380.4 | NP_759012.1 | 2 - 3 | 267 | 69 | ||
| GenBank mRNA | AK031608.1 | - | 2 - 3 | 206 | 69 | |
| AK035948.1 | - | 2 - 3 | 215 | 69 | ||
| AK036224.1 | - | 2 - 3 | 197 | 69 | ||
| AK045278.1 | - | 2 - 3 | 415 | 69 | ||
| AK079886.1 | - | 2 - 3 | 236 | 69 | ||
| AK082556.1 | - | 2 - 3 | 437 | 69 | ||
| AK167641.1 | - | 2 - 3 | 437 | 69 | ||
| AK197458.1 | - | 1 - 2 | 307 | 69 | ||
| AK215456.1 | - | 2 - 3 | 109 | 69 | ||
| BC026809.1 | - | 2 - 3 | 218 | 4205469 | 69 | |
| CB249259.1 | - | 2 - 3 | 268 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045278.1, AK167641.1, NM_001300827.1 transcripts.
Set Membership: |
|

Poglut1
Genomic Map