Assay Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300827.1 | NP_001287756.1 | 4 - 5 | 741 | 77 | |
| NM_172380.4 | NP_759012.1 | 4 - 5 | 538 | 77 | ||
| GenBank mRNA | AK031608.1 | - | 4 - 5 | 477 | 77 | |
| AK035948.1 | - | 4 - 5 | 486 | 77 | ||
| AK036224.1 | - | 4 - 5 | 468 | 77 | ||
| AK041321.1 | - | 4 - 5 | 476 | 77 | ||
| AK045278.1 | - | 4 - 5 | 686 | 77 | ||
| AK079886.1 | - | 4 - 5 | 507 | 77 | ||
| AK167641.1 | - | 4 - 5 | 708 | 77 | ||
| BC026809.1 | - | 4 - 5 | 489 | 4205469 | 77 | |
| CB249259.1 | - | 4 - 5 | 539 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Poglut1
Genomic Map