Assay Details
| Gene Symbol: Poglut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300827.1 | NP_001287756.1 | 6 - 7 | 934 | 113 | |
| NM_172380.4 | NP_759012.1 | 6 - 7 | 731 | 113 | ||
| GenBank mRNA | AK031608.1 | - | 6 - 7 | 670 | 113 | |
| AK035948.1 | - | 6 - 7 | 679 | 113 | ||
| AK036224.1 | - | 6 - 7 | 661 | 113 | ||
| AK041321.1 | - | 6 - 7 | 669 | 113 | ||
| AK045278.1 | - | 6 - 7 | 879 | 113 | ||
| AK079886.1 | - | 6 - 7 | 700 | 113 | ||
| AK167641.1 | - | 6 - 7 | 901 | 113 | ||
| BC026809.1 | - | 6 - 7 | 682 | 4205469 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poglut1
Genomic Map