Assay Details
| Gene Symbol: App | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198823.1 | NP_001185752.1 | 5 - 6 | 807 | 170 | |
| NM_001198824.1 | NP_001185753.1 | 5 - 6 | 807 | 170 | ||
| NM_001198825.1 | NP_001185754.1 | 5 - 6 | 807 | 170 | ||
| NM_001198826.1 | NP_001185755.1 | 5 - 6 | 807 | 170 | ||
| NM_007471.3 | NP_031497.2 | 5 - 6 | 807 | 170 | ||
| GenBank mRNA | AK133639.1 | - | 5 - 6 | 804 | 170 | |
| AK147543.1 | - | 5 - 6 | 843 | 170 | ||
| AK159245.1 | - | 5 - 6 | 816 | 170 | ||
| AK159352.1 | - | 5 - 6 | 814 | 170 | ||
| AK159718.1 | - | 5 - 6 | 793 | 170 | ||
| BC005490.1 | - | 1 - 2 | 171 | 170 | ||
| BC070409.1 | - | 5 - 6 | 803 | 5708093 | 170 | |
| U84012.1 | - | 5 - 6 | 796 | 170 | ||
More Information
Set Membership: |
|

App
Genomic Map