Assay Details
| Gene Symbol: Adamts5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011782.2 | NP_035912.2 | 3 - 4 | 2257 | 65 | |
| GenBank mRNA | AF140673.1 | - | 3 - 4 | 1416 | 65 | |
| AK044746.1 | - | 3 - 4 | 479 | 65 | ||
| AK082475.1 | - | 3 - 4 | 479 | 65 | ||
| BC138619.1 | - | 3 - 4 | 1441 | 8861639 | 65 | |
| BC138620.1 | - | 3 - 4 | 1441 | 8861640 | 65 | |
More Information
Set Membership: |
|

Adamts5
Genomic Map