Assay Details
| Gene Symbol: Tomm70a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138599.5 | NP_613065.2 | 2 - 3 | 655 | 81 | |
| GenBank mRNA | AK012084.1 | - | 2 - 3 | 656 | 81 | |
| AK083586.1 | - | 2 - 3 | 656 | 81 | ||
| AK122356.1 | - | 588 | 81 | |||
| AK145458.1 | - | 2 - 3 | 668 | 81 | ||
| AK150429.1 | - | 2 - 3 | 656 | 81 | ||
| AK161720.1 | - | 2 - 3 | 645 | 81 | ||
| AK196165.1 | - | 2 - 3 | 224 | 81 | ||
| BC057096.1 | - | 2 - 3 | 656 | 5709110 | 81 | |
| BC139420.1 | - | 2 - 3 | 628 | 8862442 | 81 | |
| BC139421.1 | - | 2 - 3 | 628 | 8862443 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tomm70a
Genomic Map