Assay Details
| Gene Symbol: Tbc1d23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026254.2 | NP_080530.2 | 12 - 13 | 1408 | 113 | |
| GenBank mRNA | AK015437.1 | - | 3 - 4 | 240 | 113 | |
| AK155142.1 | - | 12 - 13 | 1429 | 113 | ||
| BC019762.1 | - | 2 - 3 | 146 | 113 | ||
| BC031706.1 | - | 12 - 13 | 1408 | 4194266 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d23
Genomic Map