Assay Details
| Gene Symbol: Tmem30c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027651.1 | NP_081927.1 | 5 - 6 | 565 | 109 | |
| GenBank mRNA | AK016597.1 | - | 5 - 6 | 579 | 109 | |
| AK161475.1 | - | 4 - 5 | 2429 | 109 | ||
More Information
Set Membership: |
|

Tmem30c
Genomic Map