Assay Details
| Gene Symbol: Psmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134101.2 | NP_598862.1 | 3 - 4 | 422 | 92 | |
| NR_027485.1 | 3 - 4 | 514 | 92 | |||
| GenBank mRNA | AK146482.1 | - | 3 - 4 | 371 | 92 | |
| AK150374.1 | - | 3 - 4 | 394 | 92 | ||
| AK151588.1 | - | 3 - 4 | 394 | 92 | ||
| AK152007.1 | - | 3 - 4 | 394 | 92 | ||
| AK152970.1 | - | 3 - 4 | 391 | 92 | ||
| AK159636.1 | - | 3 - 4 | 371 | 92 | ||
| AK159756.1 | - | 3 - 4 | 394 | 92 | ||
| AK159815.1 | - | 3 - 4 | 390 | 92 | ||
| AK166816.1 | - | 3 - 4 | 374 | 92 | ||
| AK167326.1 | - | 3 - 4 | 374 | 92 | ||
| AK167774.1 | - | 3 - 4 | 371 | 92 | ||
| AK167786.1 | - | 3 - 4 | 372 | 92 | ||
| AK167909.1 | - | 3 - 4 | 382 | 92 | ||
| AK167995.1 | - | 3 - 4 | 390 | 92 | ||
| AK168131.1 | - | 3 - 4 | 381 | 92 | ||
| AK168398.1 | - | 3 - 4 | 374 | 92 | ||
| AK168543.1 | - | 3 - 4 | 371 | 92 | ||
| AK169867.1 | - | 3 - 4 | 374 | 92 | ||
| AK180397.1 | - | 407 | 92 | |||
| AK180842.1 | - | 3 - 4 | 250 | 92 | ||
| BC021508.1 | - | 3 - 4 | 391 | 5347139 | 92 | |
| BC024058.1 | - | 3 - 4 | 364 | 92 | ||
| BC024830.1 | - | 3 - 4 | 362 | 5358424 | 92 | |
| BC027388.1 | - | 3 - 4 | 386 | 4952935 | 92 | |
| BC027767.1 | - | 3 - 4 | 390 | 5347281 | 92 | |
| BC027822.1 | - | 3 - 4 | 373 | 5321505 | 92 | |
| BC028851.1 | - | 3 - 4 | 362 | 3660169 | 92 | |
| BC031128.1 | - | 3 - 4 | 391 | 4982313 | 92 | |
| BC033325.1 | - | 3 - 4 | 451 | 92 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmd2
Genomic Map