Assay Details
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039533.2 | NP_001034622.1 | 10 - 11 | 1075 | 71 | |
| NM_001291017.1 | NP_001277946.1 | 10 - 11 | 1075 | 71 | ||
| NM_053181.3 | NP_444411.2 | 10 - 11 | 1075 | 71 | ||
| GenBank mRNA | AK004611.1 | - | 10 - 11 | 952 | 71 | |
| AK030613.1 | - | 10 - 11 | 906 | 71 | ||
| AK033107.1 | - | 10 - 11 | 906 | 71 | ||
| AK054329.1 | - | 10 - 11 | 1058 | 71 | ||
| AK163696.1 | - | 10 - 11 | 946 | 71 | ||
| AK165587.1 | - | 10 - 11 | 910 | 71 | ||
| AK170661.1 | - | 10 - 11 | 1062 | 71 | ||
| AK182009.1 | - | 2 - 3 | 103 | 71 | ||
| BC005541.1 | - | 10 - 11 | 893 | 3590316 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdxdc1
Genomic Map