Assay Details
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039533.2 | NP_001034622.1 | 15 - 16 | 1504 | 78 | |
| NM_001291017.1 | NP_001277946.1 | 15 - 16 | 1504 | 78 | ||
| NM_053181.3 | NP_444411.2 | 15 - 16 | 1504 | 78 | ||
| GenBank mRNA | AK004611.1 | - | 15 - 16 | 1381 | 78 | |
| AK030613.1 | - | 15 - 16 | 1335 | 78 | ||
| AK033107.1 | - | 15 - 16 | 1335 | 78 | ||
| AK054329.1 | - | 15 - 16 | 1487 | 78 | ||
| AK075829.1 | - | 8 - 9 | 627 | 78 | ||
| AK163696.1 | - | 15 - 16 | 1375 | 78 | ||
| AK165587.1 | - | 15 - 16 | 1339 | 78 | ||
| BC005541.1 | - | 15 - 16 | 1322 | 3590316 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdxdc1
Genomic Map