Assay Details
| Gene Symbol: Pdxdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039533.2 | NP_001034622.1 | 1 - 2 | 236 | 86 | |
| NM_001291017.1 | NP_001277946.1 | 1 - 2 | 236 | 86 | ||
| NM_053181.3 | NP_444411.2 | 1 - 2 | 236 | 86 | ||
| GenBank mRNA | AK004611.1 | - | 1 - 2 | 113 | 86 | |
| AK030613.1 | - | 1 - 2 | 67 | 86 | ||
| AK033107.1 | - | 1 - 2 | 67 | 86 | ||
| AK054329.1 | - | 1 - 2 | 219 | 86 | ||
| AK163696.1 | - | 1 - 2 | 107 | 86 | ||
| AK165587.1 | - | 1 - 2 | 71 | 86 | ||
| AK170661.1 | - | 1 - 2 | 223 | 86 | ||
| BC005541.1 | - | 1 - 2 | 54 | 3590316 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK004611.1, AK030613.1, AK033107.1, AK054329.1, AK163696.1, AK165587.1, AK170661.1, BC005541.1, NM_001039533.2, NM_001291017.1, NM_053181.3 transcripts.
Set Membership: |
|

Pdxdc1
Genomic Map