Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 13 - 14 | 2903 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marf1| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 13 - 14 | 2903 | 109 | |
Set Membership: |
|