Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 21 - 22 | 4361 | 102 | |
| GenBank mRNA | AK053159.1 | - | 1 - 2 | 79 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Marf1
Genomic Map