Assay Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081154.2 | NP_001074623.1 | 4 - 5 | 1194 | 65 | |
| GenBank mRNA | AK035704.1 | - | 848 | 65 | ||
| AK220341.1 | - | 2 - 3 | 2976 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Marf1
Genomic Map