Assay Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039521.1 | NP_001034610.1 | 15 - 16 | 1604 | 74 | |
| GenBank mRNA | AK080397.1 | - | 15 - 16 | 1560 | 74 | |
| AK080680.1 | - | 1584 | 74 | |||
| AK165676.1 | - | 15 - 16 | 1592 | 74 | ||
| BC034110.1 | - | 11 - 12 | 1119 | 74 | ||
| BC055781.1 | - | 2 - 3 | 222 | 74 | ||
| BC138774.1 | - | 15 - 16 | 1569 | 8861796 | 74 | |
| BC138776.1 | - | 15 - 16 | 1569 | 8861798 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrn3
Genomic Map