Assay Details
| Gene Symbol: Parn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028761.3 | NP_083037.1 | 2 - 3 | 234 | 64 | |
| GenBank mRNA | AK004572.1 | - | 2 - 3 | 235 | 64 | |
| AK041044.1 | - | 2 - 3 | 203 | 64 | ||
| AK045181.1 | - | 2 - 3 | 220 | 64 | ||
| AK049829.1 | - | 2 - 3 | 214 | 64 | ||
| AK160611.1 | - | 2 - 3 | 203 | 64 | ||
More Information
Set Membership: |
|

Parn
Genomic Map