Assay Details
| Gene Symbol: Ncbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026554.4 | NP_080830.1 | 3 - 4 | 507 | 82 | |
| GenBank mRNA | AK008980.1 | - | 3 - 4 | 447 | 82 | |
| AK012659.1 | - | 3 - 4 | 427 | 82 | ||
| AK077841.1 | - | 3 - 4 | 415 | 82 | ||
| AK078181.1 | - | 3 - 4 | 438 | 82 | ||
| AK135597.1 | - | 3 - 4 | 422 | 82 | ||
| AK147027.1 | - | 3 - 4 | 439 | 82 | ||
| BC118926.2 | - | 3 - 4 | 415 | 40090166 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ncbp2
Genomic Map