Assay Details
| Gene Symbol: N6amt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026366.2 | NP_080642.1 | 4 - 5 | 465 | 86 | |
| GenBank mRNA | AK012019.1 | - | 4 - 5 | 405 | 86 | |
| AK145617.1 | - | 4 - 5 | 417 | 86 | ||
| AY456393.1 | - | 4 - 5 | 465 | 86 | ||
| BC116393.1 | - | 4 - 5 | 435 | 40101877 | 86 | |
| BC116394.1 | - | 4 - 5 | 435 | 40101881 | 86 | |
More Information
Set Membership: |
|

N6amt1
Genomic Map