Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_001130484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_001130484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_001130485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_001130485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_021338.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
NM_021338.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004353.mm.1 |
TC1600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004347.mm.1 |
NM_021338.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004347.mm.1 |
NM_021338.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004347.mm.1 |
TC1600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_001130484.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_001130484.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_001130485.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_001130485.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_021338.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
NM_021338.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004354.mm.1 |
TC1600000460.mm.1 |
MTA TC |