Assay Details
| Gene Symbol: Lrch3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081255.1 | NP_001074724.1 | 17 - 18 | 1907 | 61 | |
| GenBank mRNA | AK134026.1 | - | 7 - 8 | 539 | 61 | |
| AK155550.1 | - | 17 - 18 | 1905 | 61 | ||
| BC151058.1 | - | 17 - 18 | 1885 | 9087971 | 61 | |
More Information
Set Membership: |
|

Lrch3
Genomic Map