Assay Details
| Gene Symbol: Iqcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178378.3 | NP_848465.1 | 10 - 11 | 1238 | 69 | |
| GenBank mRNA | AK010269.1 | - | 7 - 8 | 787 | 69 | |
| BC049990.1 | - | 10 - 11 | 1222 | 6310844 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655998 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007356 | BC049990.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007356 | AK146422.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2007356 | NM_178378.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678770 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74499 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 103393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 801907 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 735412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246935 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 771986 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880173 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 188479 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921630 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601713 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 738070 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 703494 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 253279 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 6271 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55911 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116949 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110387 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130407 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007356 | BC049990.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007356 | AK146422.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2007356 | NM_178378.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30655990 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110387 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03164796 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130407 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08556029 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03164799 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08556106 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08556068 | - | - |
| Whole Mouse Genome G4122A | A_51_P152550 | BC049990.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152550 | AK010269.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152550 | AK146422.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P152550 | NM_178378.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110320 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03164799 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110387 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130407 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12878170 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12878163 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12878157 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12878174 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655996 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655994 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655997 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655998 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655993 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30655991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655866 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30656020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655878 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30655975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12975535 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12920337 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12878157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655971 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12878173 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12975534 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12554216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655983 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12920336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655887 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655894 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12629395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13051461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12931151 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655987 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30655979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12569296 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10439122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439118 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10439119 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460467_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10439114 | - | - |
| Mouse Genome 430 2.0 Array | 1460467_at | - | - |
| Mouse Genome 430 2.0 Array | 1460467_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69707_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69707_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600012464.mm.1 | NM_178378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012464.mm.1 | NM_178378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012464.mm.1 | TC1600001519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600012465.mm.1 | NM_178378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012465.mm.1 | NM_178378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012465.mm.1 | TC1600001519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600012466.mm.1 | NM_178378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012466.mm.1 | NM_178378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012466.mm.1 | TC1600001519.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1600012463.mm.1 | NM_178378.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012463.mm.1 | NM_178378.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600012463.mm.1 | TC1600001519.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460467_at | - | - |
| Mouse Expression Set 430 | 1460467_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460467_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1460467_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 772486 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4780059 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4780059 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004780059 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2700558 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2700558 | - | - |
| mouse-6 v1.1 (Illumina) | scl48539.12_329-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4780059 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl48539.12_329-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4780059 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241911 | - | - |

Iqcg
Genomic Map