Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 1 - 2 | 145 | 126 | |
| GenBank mRNA | AK028259.1 | - | 1 - 2 | 146 | 126 | |
| AK028307.1 | - | 1 - 2 | 164 | 126 | ||
| AK154800.1 | - | 1 - 2 | 163 | 126 | ||
| AK169311.1 | - | 1 - 2 | 168 | 126 | ||
| AK169348.1 | - | 1 - 2 | 168 | 126 | ||
| AK169464.1 | - | 1 - 2 | 184 | 126 | ||
| BC006771.1 | - | 1 - 2 | 130 | 3588534 | 126 | |
| U20619.1 | - | 1 - 2 | 148 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK028259.1, AK028307.1, AK154800.1, AK169311.1, AK169348.1, BC006771.1, NM_008465.5 transcripts.
Set Membership: |
|

Kpna1
Genomic Map