Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 2 - 3 | 273 | 81 | |
| GenBank mRNA | AK028259.1 | - | 2 - 3 | 274 | 81 | |
| AK028307.1 | - | 2 - 3 | 292 | 81 | ||
| AK154800.1 | - | 2 - 3 | 291 | 81 | ||
| AK169311.1 | - | 2 - 3 | 296 | 81 | ||
| AK169348.1 | - | 2 - 3 | 296 | 81 | ||
| AK169464.1 | - | 2 - 3 | 312 | 81 | ||
| BC006771.1 | - | 2 - 3 | 258 | 3588534 | 81 | |
| U20619.1 | - | 2 - 3 | 276 | 83 | ||
| U34228.1 | - | 1 - 2 | 122 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpna1
Genomic Map