Assay Details
| Gene Symbol: Kpna1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008465.5 | NP_032491.2 | 8 - 9 | 905 | 133 | |
| GenBank mRNA | AK028259.1 | - | 8 - 9 | 906 | 133 | |
| AK028307.1 | - | 8 - 9 | 924 | 133 | ||
| AK154800.1 | - | 8 - 9 | 923 | 133 | ||
| AK169311.1 | - | 8 - 9 | 928 | 133 | ||
| AK169348.1 | - | 8 - 9 | 928 | 133 | ||
| BC006771.1 | - | 8 - 9 | 890 | 3588534 | 133 | |
| U20619.1 | - | 8 - 9 | 910 | 133 | ||
| U34228.1 | - | 7 - 8 | 754 | 133 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kpna1
Genomic Map