Assay Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039530.3 | NP_001034619.2 | 15 - 16 | 5025 | 146 | |
| GenBank mRNA | AK220263.1 | - | 10 - 11 | 2617 | 146 | |
| BC047386.1 | - | 10 - 11 | 2125 | 146 | ||
| BC138874.1 | - | 15 - 16 | 5140 | 8861896 | 146 | |
| BC145452.1 | - | 14 - 15 | 5227 | 145 | ||
| DQ372930.1 | - | 15 - 16 | 5189 | 146 | ||
More Information
Set Membership: |
|

Parp14
Genomic Map