Assay Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039530.3 | NP_001034619.2 | 6 - 7 | 3144 | 115 | |
| GenBank mRNA | AK089685.1 | - | 7 - 8 | 3235 | 115 | |
| AK220263.1 | - | 1 - 2 | 736 | 115 | ||
| BC138874.1 | - | 6 - 7 | 3259 | 8861896 | 115 | |
| BC145452.1 | - | 6 - 7 | 3259 | 115 | ||
| DQ372930.1 | - | 6 - 7 | 3308 | 115 | ||
More Information
Set Membership: |
|

Parp14
Genomic Map