Assay Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039530.3 | NP_001034619.2 | 8 - 9 | 3606 | 101 | |
| GenBank mRNA | AK089685.1 | - | 9 - 10 | 3697 | 101 | |
| AK220263.1 | - | 3 - 4 | 1198 | 101 | ||
| BC047386.1 | - | 3 - 4 | 707 | 101 | ||
| BC138874.1 | - | 8 - 9 | 3721 | 8861896 | 101 | |
| BC145452.1 | - | 8 - 9 | 3721 | 101 | ||
| DQ372930.1 | - | 8 - 9 | 3770 | 101 | ||
More Information
Set Membership: |
|

Parp14
Genomic Map