Assay Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039530.3 | NP_001034619.2 | 10 - 11 | 3829 | 83 | |
| GenBank mRNA | AK089685.1 | - | 11 - 12 | 3920 | 83 | |
| AK220263.1 | - | 5 - 6 | 1421 | 83 | ||
| BC047386.1 | - | 5 - 6 | 930 | 83 | ||
| BC138874.1 | - | 10 - 11 | 3944 | 8861896 | 83 | |
| BC145452.1 | - | 10 - 11 | 3944 | 83 | ||
| DQ372930.1 | - | 10 - 11 | 3993 | 83 | ||
More Information
Set Membership: |
|

Parp14
Genomic Map