Assay Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039530.3 | NP_001034619.2 | 13 - 14 | 4284 | 89 | |
| GenBank mRNA | AK220263.1 | - | 8 - 9 | 1876 | 89 | |
| BC021340.1 | - | 1 - 2 | 135 | 89 | ||
| BC138874.1 | - | 13 - 14 | 4399 | 8861896 | 89 | |
| DQ372930.1 | - | 13 - 14 | 4448 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parp14
Genomic Map