Assay Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198101.1 | NP_932769.1 | 15 - 16 | 1668 | 79 | |
| GenBank mRNA | AK019874.2 | - | 4 - 5 | 418 | 79 | |
| AK052221.1 | - | 15 - 16 | 1633 | 79 | ||
| AK137177.1 | - | 13 - 14 | 1661 | 79 | ||
| BC057037.1 | - | 15 - 16 | 1668 | 5702855 | 79 | |
| BC060628.1 | - | 15 - 16 | 1643 | 6847400 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmip
Genomic Map