Assay Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198101.1 | NP_932769.1 | 5 - 6 | 492 | 68 | |
| GenBank mRNA | AK043883.1 | - | 5 - 6 | 491 | 68 | |
| AK052221.1 | - | 5 - 6 | 459 | 68 | ||
| AK137177.1 | - | 3 - 4 | 485 | 68 | ||
| AK172438.1 | - | 5 - 6 | 480 | 68 | ||
| BC057037.1 | - | 5 - 6 | 492 | 5702855 | 68 | |
| BC060628.1 | - | 5 - 6 | 467 | 6847400 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmip
Genomic Map