Assay Details
| Gene Symbol: Gmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198101.1 | NP_932769.1 | 6 - 7 | 556 | 70 | |
| GenBank mRNA | AK043883.1 | - | 6 - 7 | 555 | 70 | |
| AK052221.1 | - | 6 - 7 | 523 | 70 | ||
| AK082337.1 | - | 6 - 7 | 553 | 70 | ||
| AK137177.1 | - | 4 - 5 | 549 | 70 | ||
| AK172438.1 | - | 6 - 7 | 544 | 70 | ||
| AK216290.1 | - | 2 - 3 | 86 | 70 | ||
| BC057037.1 | - | 6 - 7 | 556 | 5702855 | 70 | |
| BC060628.1 | - | 6 - 7 | 531 | 6847400 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gmip
Genomic Map