Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK047994.1 | - | 1 - 2 | 163 | 145 | |
| AK077603.1 | - | 1 - 2 | 229 | 145 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK047994.1 transcripts.
Set Membership: |
|

Mef2c
Genomic Map