Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170537.1 | NP_001164008.1 | 4 - 5 | 833 | 58 | |
| GenBank mRNA | AK077603.1 | - | 3 - 4 | 622 | 58 | |
| AK132573.1 | - | 4 - 5 | 662 | 58 | ||
| BC026841.1 | - | 3 - 4 | 565 | 4481227 | 58 | |
| L13171.1 | - | 2 - 3 | 394 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2c
Genomic Map