Assay Details
| Gene Symbol: Tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252443.1 | NP_001239372.1 | 5 - 6 | 883 | 65 | |
| NM_019678.3 | NP_062652.1 | 6 - 7 | 1030 | 65 | ||
| GenBank mRNA | AK088864.1 | - | 5 - 6 | 701 | 65 | |
| AK158539.1 | - | 6 - 7 | 896 | 65 | ||
| AK167475.1 | - | 6 - 7 | 894 | 65 | ||
| AK194521.1 | - | 1 - 2 | 48 | 65 | ||
| BC024638.1 | - | 6 - 7 | 808 | 4015300 | 65 | |
| U94662.1 | - | 6 - 7 | 932 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tfg
Genomic Map