Assay Details
| Gene Symbol: Cox18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033310.3 | NP_001028482.2 | 3 - 4 | 648 | 72 | |
| NR_028088.1 | 3 - 4 | 652 | 72 | |||
| GenBank mRNA | AK156053.1 | - | 3 - 4 | 617 | 72 | |
| BC021612.1 | - | 1 - 2 | 199 | 72 | ||
| BC138972.1 | - | 3 - 4 | 626 | 8861994 | 72 | |
| BC145486.1 | - | 3 - 4 | 630 | 72 | ||
| BC145728.1 | - | 3 - 4 | 626 | 40131031 | 72 | |
| DV661727.1 | - | 2 - 3 | 259 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox18
Genomic Map