Assay Details
| Gene Symbol: Top3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011624.2 | NP_035754.1 | 9 - 10 | 1219 | 112 | |
| GenBank mRNA | AB013603.1 | - | 8 - 9 | 1032 | 112 | |
| AB045324.1 | - | 9 - 10 | 1293 | 112 | ||
| AK144709.1 | - | 10 - 11 | 1205 | 112 | ||
| AK146475.1 | - | 9 - 10 | 1241 | 112 | ||
| AK163796.1 | - | 2 - 3 | 228 | 112 | ||
| AK205914.1 | - | 2 - 3 | 212 | 112 | ||
| BC031723.1 | - | 9 - 10 | 1219 | 5003002 | 112 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top3b
Genomic Map