Assay Details
| Gene Symbol: Ppil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252444.1 | NP_001239373.1 | 3 - 4 | 205 | 69 | |
| NM_001252445.1 | NP_001239374.1 | 3 - 4 | 205 | 69 | ||
| NM_144954.3 | NP_659203.1 | 3 - 4 | 205 | 69 | ||
| GenBank mRNA | AK009460.1 | - | 3 - 4 | 166 | 69 | |
| AK012472.1 | - | 3 - 4 | 186 | 69 | ||
| AK081792.1 | - | 3 - 4 | 187 | 69 | ||
| AK090052.1 | - | 3 - 4 | 198 | 69 | ||
| AK161596.1 | - | 3 - 4 | 190 | 69 | ||
| BY033661.1 | - | 3 - 4 | 214 | 69 | ||
| BY145175.1 | - | 3 - 4 | 202 | 69 | ||
More Information
Set Membership: |
|

Ppil2
Genomic Map