Assay Details
| Gene Symbol: 4933404G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045920.1 | 1 - 2 | 142 | 149 | ||
| GenBank mRNA | AK016605.1 | - | 1 - 2 | 120 | 149 | |
| AK016649.1 | - | 1 - 2 | 118 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016649.1 transcripts.
Set Membership: |
|

4933404G15Rik
Genomic Map