Assay Details
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030558.2 | NP_085035.1 | 5 - 6 | 609 | 124 | |
| GenBank mRNA | AF231122.1 | - | 5 - 6 | 561 | 124 | |
| BC019975.1 | - | 5 - 6 | 609 | 4223351 | 124 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Car15
Genomic Map